Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900799

Sigma-Aldrich

Trifluoroacetic acid

Vetec, reagent grade, 99%

Synonym(s):

TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein/REAXYS Number:
742035
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

product line

Vetec

assay

99%

form

liquid

refractive index

n20/D 1.3 (lit.)

pH

1 (20 °C, 1 g/L)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

density

1.489 g/mL at 20 °C (lit.)

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 2

flash_point_f

>212.0 °F - Pensky-Martens closed cup

flash_point_c

> 100 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Plourde et al.
Lancet (London, England), 386(10009), 2192-2203 (2015-09-29)
Transradial access for cardiac catheterisation results in lower bleeding and vascular complications than the traditional transfemoral access route. However, the increased radiation exposure potentially associated with transradial access is a possible drawback of this method. Whether transradial access is associated
Wen Zhang et al.
Methods (San Diego, Calif.), 81, 50-55 (2015-02-28)
Clinical tissue samples provide valuable information for understanding human diseases. One major type of clinical tissue sample that is amenable to various kinds of analysis is fresh frozen and optimal cutting temperature (OCT)-embedded primary patient tissue. Recent advances in mass
Christian D Kelstrup et al.
Journal of proteome research, 13(12), 6187-6195 (2014-10-29)
Shotgun proteomics is a powerful technology for global analysis of proteins and their post-translational modifications. Here, we investigate the faster sequencing speed of the latest Q Exactive HF mass spectrometer, which features an ultra-high-field Orbitrap mass analyzer. Proteome coverage is
Szabolcs Fekete et al.
Journal of chromatography. A, 1393, 73-80 (2015-04-01)
The goal of this work was to evaluate the changes in retention induced by frictional heating, pressure and temperature under ultra high pressure liquid chromatography (UHPLC) conditions, for four model proteins (i.e. lysozyme, myoglobin, fligrastim and interferon alpha-2A) possessing molecular
Oskar Karlsson et al.
Archives of toxicology, 89(3), 423-436 (2014-05-07)
The environmental neurotoxin β-N-methylamino-L-alanine (BMAA) has been implicated in the etiology of neurodegenerative disease, and recent studies indicate that BMAA can be misincorporated into proteins. BMAA is a developmental neurotoxicant that can induce long-term learning and memory deficits, as well

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service