Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900794

Sigma-Aldrich

4-Hydroxybenzoic acid

Vetec, reagent grade, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2H
CAS Number:
Molecular Weight:
138.12
Beilstein/REAXYS Number:
970950
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

99%

mp

213-217 °C (lit.)

SMILES string

OC(=O)c1ccc(O)cc1

InChI

1S/C7H6O3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H,(H,9,10)

InChI key

FJKROLUGYXJWQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M G Soni et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 39(6), 513-532 (2001-05-11)
Propyl paraben (CAS no. 94-13-3) is a stable, non-volatile compound used as an antimicrobial preservative in foods, drugs and cosmetics for over 50 years. It is an ester of p-hydroxybenzoate. Propyl paraben is readily absorbed via the gastrointestinal tract and
L C Goudas et al.
Anesthesia and analgesia, 89(5), 1209-1215 (1999-11-30)
Intracerebroventricular (ICV) morphine administration is effective for the management of refractory cancer pain. Recent preclinical observations of acute depletion of the major endogenous intracellular antioxidant glutathione (GSH) in brain and peripheral organs after ICV morphine in rodents led us to
Letian X Xie et al.
The Journal of biological chemistry, 287(28), 23571-23581 (2012-05-18)
Most of the Coq proteins involved in coenzyme Q (ubiquinone or Q) biosynthesis are interdependent within a multiprotein complex in the yeast Saccharomyces cerevisiae. Lack of only one Coq polypeptide, as in Δcoq strains, results in the degradation of several
Suri Roowi et al.
Molecular nutrition & food research, 53 Suppl 1, S68-S75 (2009-05-06)
Human urine was collected over a 24 h period after the consumption of 250 mL of (i) water, (ii) orange juice, and (iii) orange juice plus 150 mL of full fat natural yoghurt. The orange juice contained 168 micromol of
L I Wiebe et al.
Drug metabolism and disposition: the biological fate of chemicals, 6(3), 296-302 (1978-05-01)
Butylated hydroxytoluene (BHT) containing the stable isotope 13C was synthesized from 2-[13C]methylpropan-2-ol. A minor constituent of urine following ingestion of BHT-13C by a human volunteer was identified as 3,5-di-(1-[13C]methyl-1-methylethyl)-4-hydroxybenzoic acid, The major metabolite detected was 13C-labeled 5-carboxy-7-(1-carboxy-1-methylethyl)-3,3-dimethyl-2-hydroxy-2,3-dihydrobenzofuran. Detailed spectral analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service