Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900792

Sigma-Aldrich

3-Nitrobenzaldehyde

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CHO
CAS Number:
Molecular Weight:
151.12
Beilstein/REAXYS Number:
386795
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

55-58 °C (lit.)

SMILES string

[O-][N+](=O)c1cccc(C=O)c1

InChI

1S/C7H5NO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-5H

InChI key

ZETIVVHRRQLWFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Karunakaran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 98, 229-239 (2012-09-11)
FT-IR (4000-400 cm(-1)) and FT-Raman (3500-100 cm(-1)) spectral measurements of 4-chloro-3-nitrobenzaldehyde have been done. Ab initio (HF/6-311+G(d,p)) and DFT (B3LYP/6-311+G(d,p)) calculations have been performed giving energies, optimized structures, harmonic vibrational frequencies, infrared intensities and Raman activities. A detailed interpretation of
E V Sargent et al.
Mutation research, 175(3), 133-137 (1986-11-01)
m-Nitrobenzaldehyde (MNB) was evaluated for mutagenic activity using the Ames microbial mutagenicity test and for its ability to induce DNA single-strand breaks in rat hepatocytes as measured by alkaline elution. MNB was tested in S. typhimurium strains TA1535, TA1537, TA1538
Halil Gökce et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 79(5), 1783-1793 (2011-06-21)
The molecular geometry, vibrational frequencies, 1H and 13C NMR chemical shifts, UV-vis spectra, HOMO-LUMO analyses, molecular electrostatic potentials (MEPs), , thermodynamic properties and atomic charges of 3- and 4-Nitrobenzaldehyde oxime (C7H6N2O3) molecules have been investigated by using Hartree-Fock (HF) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service