Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900788

Sigma-Aldrich

3-Aminobenzoic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Beilstein/REAXYS Number:
471603
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

reaction suitability

reaction type: solution phase peptide synthesis

color

white to brown

mp

178-180 °C (lit.)

SMILES string

Nc1cccc(c1)C(O)=O

InChI

1S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H,9,10)

InChI key

XFDUHJPVQKIXHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Yamamoto et al.
Journal of pharmaceutical sciences, 80(11), 1067-1071 (1991-11-01)
Absorption and metabolic characteristics of p-aminobenzoic acid (PABA) and m-aminobenzoic acid (MABA) from the rat small intestine were examined by means of in situ recirculation and in vitro everted sac experiments. p-Aminobenzoic acid was extremely rapidly absorbed from the rat
U Altenschmidt et al.
European journal of biochemistry, 194(2), 647-653 (1990-12-12)
A new pathway for aerobic metabolism of 2-aminobenzoate which proceeds via anthranoyl-CoA has recently been revealed in a Pseudomonas strain KB740. This bacterial strain was found to contain a small 8.1-kbp plasmid pKB740 which appears to harbour the genes encoding
Satoru Kamoda et al.
Journal of chromatography. A, 1050(2), 211-216 (2004-10-29)
Carbohydrate chains in glycoprotein pharmaceuticals have important roles for the expression of their biological activities. Therefore, development of an assessment method for the carbohydrate chains is an important parameter for quality control of glycoprotein pharmaceuticals such as newly developed therapeutic
M Nagao et al.
Environmental health perspectives, 93, 169-174 (1991-06-01)
A poly(ADP-ribose) polymerase inhibitor, benzamide (BA), was found to induce flat revertants of NIH 3T3 cells that had been transformed by human Ha-ras, rat Ki-ras, rat c-raf, and human ret-II. These genes had been amplified in original transformants, but they
Seok Min Yoon et al.
Langmuir : the ACS journal of surfaces and colloids, 23(23), 11875-11882 (2007-10-09)
One-dimensional (1D) helical organic nanostructures were synthesized by a modified vapor-solid (VS) process, called the vaporization-condensation-recrystallization (VCR) process. The conventional solution-phase synthetic methods generally mediate self-assemblies of repeating unit molecules. To provide enough intermolecular interaction forces among the unit molecules

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service