Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900787

Sigma-Aldrich

m-Toluic acid

Vetec, reagent grade, 98%

Synonym(s):

3-Methylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CO2H
CAS Number:
Molecular Weight:
136.15
Beilstein/REAXYS Number:
970526
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

263 °C (lit.)

mp

107-113 °C (lit.)

density

1.054 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)C(O)=O

InChI

1S/C8H8O2/c1-6-3-2-4-7(5-6)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

GPSDUZXPYCFOSQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Cebolla et al.
Applied and environmental microbiology, 62(1), 214-220 (1996-01-01)
A simple procedure to detect the switching on and off of catabolic promoters of Pseudomonas putida, at the level of single cells based on the immunodetection of a reporter epitope expressed on the surface of bacterial cells, has been developed.
M A Kivisaar et al.
Journal of bacteriology, 171(9), 5111-5116 (1989-09-01)
The utilization of phenol, m-toluate, and salicylate (Phe+, mTol+, and Sal+ characters, respectively) in Pseudomonas sp. strain EST1001 is determined by the coordinated expression of genes placed in different plasmids, i.e., by a multiplasmid system. The natural multiplasmid strain EST1001
K Miura et al.
Biochemistry and molecular biology international, 46(5), 933-941 (1998-12-23)
The transcription of OP2 encoding enzymes for m-toluate catabolism on the Pseudomonas putida TOL plasmid is activated by basal-level XylS protein in the presence of m-toluate or by overproduced XylS protein in the absence of m-toluate. In this study, in
C J Krieger et al.
Journal of bacteriology, 181(20), 6403-6410 (1999-10-09)
The initial enzymatic steps in anaerobic m-xylene oxidation were studied in Azoarcus sp. strain T, a denitrifying bacterium capable of mineralizing m-xylene via 3-methylbenzoate. Permeabilized cells of m-xylene-grown Azoarcus sp. strain T catalyzed the addition of m-xylene to fumarate to
M T Gallegos et al.
Journal of bacteriology, 178(8), 2356-2361 (1996-04-01)
Growth of Pseudomonas putida (pWWO) on alkylbenzoates requires the expression of the meta pathway operon, which is mediated by the XylS protein after binding of a benzoate effector. Alternatively, in cells growing on toluene or its aromatic alcohols, overexpression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service