Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900786

Sigma-Aldrich

2-Picolinic acid

Vetec, reagent grade, 98%

Synonym(s):

α-Picolinic acid, Pyridine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein/REAXYS Number:
109595
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

139-142 °C (lit.)

SMILES string

OC(=O)c1ccccn1

InChI

1S/C6H5NO2/c8-6(9)5-3-1-2-4-7-5/h1-4H,(H,8,9)

Inchi Key

SIOXPEMLGUPBBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadwiga Turło et al.
Enzyme and microbial technology, 51(6-7), 388-395 (2012-10-09)
In this report the optimization of biosynthesis of tacrolimus, the immunosupressant widely used in transplantology and dermatology was described. The enhancement of the productivity of Streptomyces tsukubaensis strain was achieved by development of new precursors of tacrolimus biosynthesis, which should
G Melillo et al.
Journal of immunology (Baltimore, Md. : 1950), 150(9), 4031-4040 (1993-05-01)
In this study we investigated the effects of picolinic acid, a catabolite of L-tryptophan, on the production of L-arginine-derived reactive nitrogen intermediates in the murine macrophage cell line ANA-1. ANA-1 macrophages did not produce nitrite (NO2-) constitutively, but accumulated detectable
Maria Carla Bosco et al.
Advances in experimental medicine and biology, 527, 55-65 (2004-06-23)
Recent studies have suggested a role for aminoacid catabolites as important regulators of macrophage (Mphi) activities. We reported previously that picolinic acid (PA), a tryptophan catabolite produced under inflammatory conditions and a costimulus with IFNgamma of Mphi effector functions, is
Haiyan An et al.
Dalton transactions (Cambridge, England : 2003), 41(27), 8390-8400 (2012-05-30)
Six 3D architectures based on lanthanide-substituted polyoxometalosilicates, KLn[(H(2)O)(6)Ln](2)[(H(2)O)(4)LnSiW(11)O(39)](2)·nH(2)O (Ln = La 1, n = 42; Ce 2, n = 40), H[(H(2)O)(6)Nd](2)[(H(2)O)(7)Nd][(H(2)O)(4)NdSiW(11)O(39)][(H(2)O)(3)NdSiW(11)O(39)]·13H(2)O (3), H(2)K(2)[(Hpic)(H(2)O)(5)Ln](2)[(H(2)O)(4)LnSiW(11)O(39)](2)·nH(2)O (Ln = La 4, n = 18.5; Ce 5, n = 35; Nd 6, n = 36;
Sarah E Coggan et al.
Journal of neurochemistry, 108(5), 1220-1225 (2009-01-15)
It has been suggested that picolinic acid (PIC), an endogenous metabolite of l-tryptophan, possesses neuro-protective and anti-proliferative effects within the CNS. However, the literature surrounding PIC is limited, and its exact endogenous function is not known. Picolinic acid is produced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service