Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900785

Sigma-Aldrich

Benzenesulfonamide

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO2NH2
CAS Number:
Molecular Weight:
157.19
Beilstein/REAXYS Number:
1100566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

149-152 °C (lit.)

SMILES string

NS(=O)(=O)c1ccccc1

InChI

1S/C6H7NO2S/c7-10(8,9)6-4-2-1-3-5-6/h1-5H,(H2,7,8,9)

InChI key

KHBQMWCZKVMBLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Zhu et al.
PloS one, 6(12), e29120-e29120 (2011-12-24)
Hemagglutinin (HA) of the influenza virus plays a crucial role in the early stage of the viral life cycle by binding to sialic acid on the surface of host epithelial cells and mediating fusion between virus envelope and endosome membrane
Asha Chandran et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 92, 84-90 (2012-03-27)
The FT-IR and FT-Raman spectra of (E)-N-carbamimidoyl-4-((4-methoxybenzylidene)amino)benzenesulfonamide were recorded and analyzed. Geometry and harmonic vibrational wavenumbers were calculated theoretically using Gaussian 03 set of quantum chemistry codes. Calculations were performed at the Hartree-Fock (HF) and density functional theory (DFT) levels
Astrid Goubet et al.
Bioorganic & medicinal chemistry letters, 23(3), 761-763 (2012-12-26)
C5-Ethynylbenzenesulfonamide-modified nucleotide (EBNA) was investigated as substrate of various DNA polymerases. The experiments revealed that KOD, Phusion and Klenow DNA polymerases successfully accepted EBNA-T nucleotide as a substrate and yielded the fully extended DNA. KOD DNA polymerase was found to
Mostafa M Ghorab et al.
Acta pharmaceutica (Zagreb, Croatia), 61(4), 415-425 (2011-12-29)
Sulfonamides and quinoxaline derivatives possess many types of biological activities and have been recently reported to show substantial antitumor activity. This paper reports the synthesis of novel thioureido sulfaquinoxaline derivatives. All the newly synthesized compounds were evaluated for their in
Edita Čapkauskaitė et al.
European journal of medicinal chemistry, 51, 259-270 (2012-03-24)
A series of [(2-pyrimidinylthio)acetyl]benzenesulfonamides were designed and synthesized. Their binding affinities as inhibitors of several recombinant human carbonic anhydrase (CA) isozymes were determined by isothermal titration calorimetry (ITC) and thermal shift assay (TSA). A group of compounds containing a chlorine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service