Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900784

Sigma-Aldrich

L-(−)-Malic acid

Vetec, reagent grade, 97%

Synonym(s):

(S)-(−)-2-Hydroxysuccinic acid, L-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
Beilstein/REAXYS Number:
1723541
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

97%

form

powder

mp

101-103 °C (lit.)

SMILES string

O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

BJEPYKJPYRNKOW-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Malic acid is a C4 dicarboxylic acid generally used as an acidulant and flavor enhancer in food industries. It is also used in pharmaceuticals, textile finishing, metal cleaning, and in the synthesis of polymalic acid, a biodegradable polymer.

Application

L-(−)-Malic acid can be used as:
  • A ligand for the synthesis of chiral triorganotin(IV) complexes.
  • A substrate in the 60Co γ-irradiation studies using electron paramagnetic resonance (EPR) technique.
  • A ligand in the complex formation studies with W(VI) using polarimeter and NMR.
  • A starting material for the preparation of organic-inorganic hybrid material named malic acid-layered double hydroxide (MA-LDH).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral self-assembly of triorganotin complexes: Syntheses, characterization, crystal structures and antitumor activity of organotin (IV) complexes containing (R)-(+)-methylsuccinic acid,(S)-(+)-methylglutaric acid and l-(−)-malic acid ligands
Ma C, et al.
Polyhedron, 31(1), 478-485 (2012)
Investigation of 60Co gamma-irradiated l-(−) malic acid, N-methyl-dl-valine and l-glutamic acid gamma-ethyl ester by electron paramagnetic resonance technique
Baskan MH, et al.
Journal of Molecular Structure, 983(1-3), 200-202 (2010)
Synthesis and structural characterization of L-(−)-malic acid pillared layered double hydroxides
Jiao FP, et al.
Latin American Applied Research, 39(2), 127-130 (2009)
Production of polymalic acid and malic acid by Aureobasidium pullulans fermentation and acid hydrolysis
Zou X, et al.
Biotechnology and Bioengineering, 110(8), 2105-2113 (2013)
Compounds of W (VI) with l (−) malic acid: a polarimetric and 1H nuclear magnetic resonance study of the formation and interconversion equilibria in excess of W (VI)
Cervilla A, et al.
Canadian Journal of Chemistry, 63(5), 1041-1047 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service