Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900780

Sigma-Aldrich

Benzoic anhydride

Vetec, reagent grade, 89%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O
CAS Number:
Molecular Weight:
226.23
Beilstein/REAXYS Number:
516726
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

89%

mp

38-42 °C (lit.)

density

1.199 g/mL at 25 °C (lit.)

SMILES string

O=C(OC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O3/c15-13(11-7-3-1-4-8-11)17-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

CHIHQLCVLOXUJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation

target_organs

Lungs

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W J Gelsema et al.
Journal of lipid research, 37(6), 1224-1233 (1996-06-01)
Benzoolysis experiments are reported in which diacylglycerophosphocholine is heated at 100 degrees C with benzoic anhydride for variable periods of time. It is shown that more than 90% of the phospholipid is dephosphorylated after 5 h of heating. Lipid extracts
J H Xu et al.
Biotechnology and bioengineering, 73(6), 493-499 (2001-05-10)
Optically active (R)-alpha-monobenzoyl glycerol (MBG) was synthesized by Candida antarctica lipase B (CHIRAZYME L-2) catalyzed asymmetric esterification of glycerol with benzoic anhydride in organic solvents. Various conditions, such as the type and composition of the organic solvent, water content of
Isamu Shiina et al.
The Journal of organic chemistry, 69(6), 1822-1830 (2004-04-03)
Various carboxylic esters are obtained at room temperature in excellent yields with high chemoselectivities from nearly equimolar amounts of carboxylic acids and alcohols using 2-methyl-6-nitrobenzoic anhydride with triethylamine by the promotion of a basic catalyst such as 4-(dimethylamino)pyridine. A variety
W J Gelsema et al.
Journal of lipid research, 38(8), 1680-1684 (1997-08-01)
Series of benzoolysis experiments with 1,2-dipalmitoyl glycerophosphocholine (GPC), 1-palmitoyl-2-lyso GPC, and 1-O-hexadecenyl-2-oleoyl GPC, as well as a few experiments with 1-oleoyl-2-acetyl GPC and 1-O-hexadecyl-2-acetyl GPC are reported. The results are used to present a consistent picture of the various dephosphorylation
P Chaturvedi et al.
Analytical biochemistry, 251(1), 89-97 (1997-08-15)
Human milk is rich in oligosaccharides, some of which inhibit toxins and pathogens involved in diseases of infants. To investigate qualitative and quantitative individual variation of human milk oligosaccharides, a sensitive method for routine identification and quantification of intact milk

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service