Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900779

Sigma-Aldrich

Ethyl benzoate

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOC2H5
CAS Number:
Molecular Weight:
150.17
Beilstein/REAXYS Number:
1908172
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

5.17 (vs air)

vapor pressure

1 mmHg ( 44 °C)

product line

Vetec

assay

98%

autoignition temp.

914 °F

refractive index

n20/D 1.504 (lit.)

bp

212 °C (lit.)

mp

−34 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)c1ccccc1

InChI

1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

MTZQAGJQAFMTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

183.2 °F - closed cup

flash_point_c

84 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Vinagre et al.
The journal of adhesive dentistry, 17(2), 107-116 (2015-04-29)
This study compared the microtensile bond strengths (μTBS) of two etch-and-rinse (ER) (OptiBond FL [OBFL]; Prime & Bond NT [PBNT]) and three self-etching (SE) (Clearfil SE Bond [CSEB]; Xeno III [XIII]; Xeno V+ [XV+]) adhesives systems to bur-prepared human enamel
Katarzyna Niedojadło et al.
Plant cell reports, 34(12), 2201-2215 (2015-09-12)
Calreticulin expression is upregulated during sexual reproduction of Hyacinthus orientalis, and the protein is localized both in the cytoplasm and a highly specialized cell wall within the female gametophyte. Several evidences indicate calreticulin (CRT) as an important calcium (Ca(2+))-binding protein
Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1405, 178-187 (2015-06-15)
The effect of three phosphonium and imidazolium ionic liquids (ILs) on the interaction between liposomes and common pharmaceuticals found in wastewaters was studied. The liposomes comprised zwitterionic phosphatidyl choline and negatively charged phosphatidyl glycerol. A set of common cationic, anionic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service