Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900777

Sigma-Aldrich

Homoveratric acid

Vetec, reagent grade, 98%

Synonym(s):

3,4-Dimethoxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
196.20
Beilstein/REAXYS Number:
1110282
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

96-98 °C (lit.)

SMILES string

COc1ccc(CC(O)=O)cc1OC

InChI

1S/C10H12O4/c1-13-8-4-3-7(6-10(11)12)5-9(8)14-2/h3-5H,6H2,1-2H3,(H,11,12)

InChI key

WUAXWQRULBZETB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[ON THE EFFECT OF NIKETHAMIDE AND HOMOVERATRIC ACID DIMETHYLAMIDE ON RESPIRATION IN INFANTS].
J PROENCA et al.
Monatsschrift fur Kinderheilkunde, 113, 341-343 (1965-04-01)
C G Charlton et al.
Life sciences, 66(22), 2159-2171 (2000-06-02)
L-dopa is the major treatment for Parkinson's disease (PD), but its efficacy is limited by the presence of dyskinesia. The dyskinesia develops over a period of exposure to L-dopa and is related to the dosage, therefore, the cause may involve
Excretion of 3,4-dimethoxyphenethylamine (DMPEA) by acute schizophrenics and controls.
A J Friedhoff et al.
Biological psychiatry, 12(5), 643-654 (1977-10-01)
K Hempel et al.
Biological psychiatry, 17(1), 49-59 (1982-01-01)
A quantitative method for the detection of DMPEA in urine was developed. It is based on the fluorometric determination of DMPEA in the form of its phosphopyridoxyl derivate. The limit of detection is 2 microgram DMPEA per 1 g creatinine.
Wai Mun Loke et al.
The Journal of nutrition, 139(12), 2309-2314 (2009-10-09)
Flavonoids are phytochemicals that are widespread in the human diet. Despite limitations in their bioavailability, experimental and epidemiological data suggest health benefits of flavonoid consumption. Valid biomarkers of flavonoid intake may be useful for estimating exposure in a range of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service