Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900776

Sigma-Aldrich

Umbelliferone

Vetec, reagent grade, 98%

Synonym(s):

7-Hydroxy-2H-1-benzopyran-2-one, 7-Hydroxycoumarin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O3
CAS Number:
Molecular Weight:
162.14
Beilstein/REAXYS Number:
127683
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

230 °C (dec.) (lit.)

SMILES string

Oc1ccc2C=CC(=O)Oc2c1

InChI

1S/C9H6O3/c10-7-3-1-6-2-4-9(11)12-8(6)5-7/h1-5,10H

InChI key

ORHBXUUXSCNDEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromu Kashida et al.
Bioorganic & medicinal chemistry, 20(14), 4310-4315 (2012-06-19)
In this study, we synthesized a simple but efficient quencher-free molecular beacon tethering 7-hydroxycoumarin on D-threoninol based on its pK(a) change. The pK(a) of 7-hydroxycoumarin in a single strand was determined as 8.8, whereas that intercalated in the duplex was
Kalyan K Sadhu et al.
Organic & biomolecular chemistry, 11(4), 563-568 (2012-10-13)
The design and synthesis of molecular probes competent for pH signaling within or beyond a certain range is a complicated matter. Herein a new mechanism for ''OFF-ON-OFF'' absorbance and fluorescence intensities vs. pH behaviour is described. The probe design is
Ye V Liu et al.
Vaccine, 33(18), 2152-2158 (2015-03-17)
In March 2013, diagnosis of the first reported case of human infection with a novel avian-origin influenza A(H7N9) virus occurred in eastern China. Most human cases have resulted in severe respiratory illness and, in some instances, death. Currently there are
Juri M Timonen et al.
European journal of medicinal chemistry, 46(9), 3845-3850 (2011-06-18)
A number of 7-hydroxycoumarins have been synthesised by Pechmann cyclisation using differently substituted resorcinols employing perchloric acid as the condensing agent. All the compounds have been characterised by analytical and spectroscopic methods. The anti-inflammatory properties were tested with LPS-induced inflammation
Seitaro Matsumoto et al.
Phytochemistry, 74, 49-57 (2011-12-16)
Ortho-hydroxylation of cinnamates is a key step in coumarin biosynthesis in plants. Ortho-hydroxylated cinnamates undergo trans/cis isomerization of the side-chain and then lactonization to form coumarins. Sweet potato [Ipomoea batatas (L.) Lam.] accumulates umbelliferone and scopoletin after biotic and abiotic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service