Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900774

Sigma-Aldrich

4,4′-Dihydroxybiphenyl

Vetec, reagent grade, 97%

Synonym(s):

4,4′-Biphenol, 4,4′-Biphenyldiol, 4,4′-Diphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4C6H4OH
CAS Number:
Molecular Weight:
186.21
Beilstein/REAXYS Number:
1908886
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

280-282 °C (lit.)

SMILES string

Oc1ccc(cc1)-c2ccc(O)cc2

InChI

1S/C12H10O2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,13-14H

InChI key

VCCBEIPGXKNHFW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 3 - Self-heat. 2 - Skin Sens. 1

Storage Class

4.2 - Pyrophoric and self-heating hazardous materials

wgk_germany

WGK 2

flash_point_f

537.8 °F

flash_point_c

281 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Golbeck et al.
Journal of bacteriology, 156(1), 49-57 (1983-10-01)
Biphenyl metabolism in Aspergillus toxicarius occurs by successive hydroxylations in the 4- and 4'-positions, followed by conjugation with sulfate to produce 4-hydroxybiphenyl-O-sulfonic acid and 4,4'-dihydroxybiphenyl-O-sulfonic acid. The hydroxylation reactions normally occur only after a prolonged lag period after which the
Jae Kyung No et al.
Biological & pharmaceutical bulletin, 29(1), 14-16 (2006-01-06)
In our previous study, we showed that 4,4'-dihydroxybiphenyl (44'-BP) reduced melanin content via the inhibition of tyrosinase. In the current study, we utilized 44'-BP treated B16 melanoma cells (B16 cells) to measure several key cellular parameters known to be involved
Rainer-B Volk et al.
Microbiological research, 161(2), 180-186 (2006-01-24)
The antimicrobial activity of two cyanobacterial exometabolites, norharmane (9H-pyrido(3,4-b)indole) and 4,4'-dihydroxybiphenyl, was determined in suspension assays. Good anticyanobacterial activities (concentrations of 8-80 microg ml(-1)) and moderate antibacterial (16-160 microg ml(-1)) and antifungal (32-40 microg ml(-1)) activities were found for both
L G McGirr et al.
Chemico-biological interactions, 60(1), 85-99 (1986-10-15)
Oxidation of p,p'-biphenol with horseradish peroxidase (HRP)-hydrogen peroxide in the presence of bovine serum albumin or with bone marrow cell homogenate-hydrogen peroxide resulted in the formation of reactive products that conjugate with protein. Glutathione prevented the protein binding. Glutathione readily
M Gill et al.
Zeitschrift fur Naturforschung. Section C, Biosciences, 39(11-12), 1027-1029 (1984-11-01)
Extraction of fresh sporophores of the fungus Agaricus xanthodermus yields 4,4'-dihydroxy-azobenzene, phenol, p-quinol, and 4,4'-dihydroxybiphenyl. This is the first report of an azo compound arising endogenously in nature, while phenol, p-quinol and 4,4'-dihydroxybiphenyl have not previously been isolated from higher

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service