Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900773

Sigma-Aldrich

Phenothiazine

Vetec, reagent grade, 98%

Synonym(s):

10H-Phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NS
CAS Number:
Molecular Weight:
199.27
Beilstein/REAXYS Number:
143237
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

bp

371 °C (lit.)

mp

182-187 °C (lit.)

SMILES string

N1c2ccccc2Sc3ccccc13

InChI

1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

WJFKNYWRSNBZNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Blood

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caiwen Ou et al.
Chemical communications (Cambridge, England), 49(18), 1853-1855 (2013-01-31)
We report a 'super-gelator' of a short peptide capped with phenothiazine acetic acid.
Alina N Sekretaryova et al.
Analytical chemistry, 84(3), 1220-1223 (2011-12-31)
Wiring glucose oxidase in the membrane with an immobilized mediator is possible due to the diffusion ability of the latter, if the enzyme containing membrane is formed according to the proposed protocol, including exposing proteins to water-organic mixtures with the
Gabriela B Rodrigues et al.
Journal of photochemistry and photobiology. B, Biology, 116, 89-94 (2012-09-18)
Photodynamic antimicrobial chemotherapy (PACT) is a promising alternative to conventional chemotherapy that can be used to treat localized mycosis. The development of PACT depends on identifying effective and selective PS for the different pathogenic species. The in vitro susceptibilities of
M L Buck et al.
DICP : the annals of pharmacotherapy, 25(3), 244-247 (1991-03-01)
A two-month-old white girl presented to our facility with increasing lethargy and new onset apnea and bradycardia following a week of upper respiratory tract infection symptoms. The patient had been receiving a cough syrup containing promethazine hydrochloride during the previous
Dalila Belei et al.
Bioorganic & medicinal chemistry letters, 22(14), 4517-4522 (2012-06-29)
The biological screening of the chemical library of our Organic Chemistry Department, carried out on an automated fluorescence-based FTase assay, allowed us to discover that a phenothiazine derivative (1d) was an inhibitor of farnesyltransferase. Three new series of human farnesyltransferase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service