Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900772

Sigma-Aldrich

Ethoxyquin

Vetec, reagent grade, 75%

Synonym(s):

1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO
CAS Number:
Molecular Weight:
217.31
Beilstein/REAXYS Number:
158223
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

75%

density

1.03 g/mL at 20 °C (lit.)

SMILES string

CCOc1ccc2NC(C)(C)C=C(C)c2c1

InChI

1S/C14H19NO/c1-5-16-11-6-7-13-12(8-11)10(2)9-14(3,4)15-13/h6-9,15H,5H2,1-4H3

InChI key

DECIPOUIJURFOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rex Munday et al.
Chemico-biological interactions, 155(3), 140-147 (2005-07-28)
Reduction of naphthoquinones by DT-diaphorase is often described as a detoxification reaction. This is true for some naphthoquinone derivatives, such as alkyl and di-alkyl naphthoquinones, but the situation with other substances, such as 2-hydroxy-1,4-naphthoquinone, is more complex. In the present
Alina Błaszczyk et al.
Chemico-biological interactions, 162(1), 70-80 (2006-06-27)
2,2,4,7-Tetramethyl-1,2,3,4-tetrahydroquinoline (THQ) is a new synthetic compound with potential antioxidant activity. In this study, cytotoxic, genotoxic and antioxidant activities of THQ were studied on human lymphocytes with the use of the trypan blue exclusion assay, the TUNEL method, the comet
J Andrew Keightley et al.
Molecular & cellular proteomics : MCP, 3(2), 167-175 (2003-12-17)
We are using a proteomic approach that combines two-dimensional electrophoresis and tandem mass spectrometry to detect and identify proteins that are differentially expressed in a cell line that is resistant to oxidative stress. The resistant cell line (OC14 cells) was
Alina Błaszczyk et al.
Cellular & molecular biology letters, 10(1), 15-21 (2005-04-06)
In our study, we analyzed the cytotoxicity of ethoxyquin (EQ) and its two salts, ethoxyquin hydrochloride (EQ-HCL) and ethoxyquin phosphate (EQ-P). It was shown that EQ was the most cytotoxic compound (IC(50) = 0.09 mM), while the lowest cytotoxic effect
Alina Błaszczyk et al.
Acta poloniae pharmaceutica, 62(2), 111-115 (2005-09-16)
In our study ethoxyquin (EQ) and its two complexes with flavonoids were obtained from ethoxyquin (1,2-dihydro-6-ethoxy-2,2,4-trimethylquinoline, EQ) and quercetin (EQ-Q, 1:1) or rutin (EQ-R, 1:1). Cytotoxicity of the tested compounds was studied using the trypan blue exclusion method and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service