Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900771

Sigma-Aldrich

1,2-Dimethoxybenzene

Vetec, reagent grade, 98%

Synonym(s):

Pyrocatechol dimethyl ether, Veratrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
Beilstein/REAXYS Number:
1364621
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.533 (lit.)

bp

206-207 °C (lit.)

mp

15 °C (lit.)

density

1.084 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1OC

InChI

1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3

Inchi Key

ABDKAPXRBAPSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

161.6 °F - closed cup

flash_point_c

72 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umma Salma Jhumur et al.
Journal of medical entomology, 43(6), 1164-1170 (2006-12-14)
Flower odors are important signals for chemical communication between plants and flower visitors. Here, we studied the naive responses of Culex pipiens pipiens biotype molestus Forskal 1775 (Diptera: Culicidae) to typical flower odors and assessed the learning capacity of mosquitoes
Y Azuma et al.
Journal of dental research, 65(1), 53-56 (1986-01-01)
The mechanism of the anti-inflammatory action of phenolic compounds was examined using neutrophil chemotaxis. Chemotactic activity of guinea pig peritoneal neutrophils to N-formylmethionyl-leucylphenylalanine (FMLP) was suppressed in a concentration-dependent manner. The order of drug potency in inhibiting the neutrophil chemotaxis
D Cai et al.
The Journal of biological chemistry, 267(16), 11149-11155 (1992-06-05)
The present study characterizes the serial reactions of H2O2 with compounds I and II of lignin peroxidase isozyme H1. These two reactions constitute part of the pathway leading to formation of the oxy complex (compound III) from the ferric enzyme.
Yoshiya Inokuchi et al.
Physical chemistry chemical physics : PCCP, 14(13), 4457-4462 (2012-02-23)
We report UV photodissociation (UVPD) and IR-UV double-resonance spectra of 1,2-dimethoxybenzene (DMB) complexes with alkali metal ions, M(+)·DMB (M = Li, Na, K, Rb, and Cs), in a cold, 22-pole ion trap. The UVPD spectrum of the Li(+) complex shows
The lowest excited triplet states of codeine and morphine and of their molecular subunits veratrole and guaiacol.
H Slama et al.
Photochemistry and photobiology, 47(5), 661-667 (1988-05-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service