Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900769

Sigma-Aldrich

Guaiacol

Vetec, reagent grade, 98%

Synonym(s):

2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)C6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein/REAXYS Number:
508112
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
grade:
reagent grade
Pricing and availability is not currently available.

grade

reagent grade

vapor density

4.27 (vs air)

vapor pressure

0.11 mmHg ( 25 °C)

product line

Vetec

assay

98%

refractive index

n20/D 1.543 (lit.)

bp

205 °C (lit.)

mp

26-29 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1O

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Guaiacol, along with 2,4,6-trichloroanisole, is responsible for cork taint in wine. A method has been developed for extraction and quantitation of the two compounds.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

179.6 °F - closed cup

flash_point_c

82 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joel D Mainland et al.
Nature neuroscience, 17(1), 114-120 (2013-12-10)
Humans have ~400 intact odorant receptors, but each individual has a unique set of genetic variations that lead to variation in olfactory perception. We used a heterologous assay to determine how often genetic polymorphisms in odorant receptors alter receptor function.
Feng Li et al.
Food chemistry, 135(2), 332-337 (2012-08-08)
Hexahydrocurcumin, 1-dehydro-[6]-gingerdione, 6-dehydroshogaol and 6-shogaol were evaluated for their antioxidant and anti-inflammatory activities in the present study. The relative antioxidant potencies of ginger compounds decreased in similar order of 1-dehydro-[6]-gingerdione, hexahydrocurcumin>6-shogaol>6-dehydroshogaol in both 1,1-diphenyl-2-picyrlhydrazyl (DPPH) radical-scavenging and trolox equivalent antioxidant
Chien-Yun Hsiang et al.
Food chemistry, 136(1), 170-177 (2012-09-29)
Ginger is a commonly used spice with anti-inflammatory potential. Colitis is the common pathological lesion of inflammatory bowel diseases. In this study, we investigated the therapeutic effects of ginger and its component zingerone in mice with 2,4,6-trinitrobenzene sulphonic acid (TNBS)-induced
Igor Cesarino et al.
Plant physiology and biochemistry : PPB, 62, 1-10 (2012-11-20)
Secreted class III peroxidases (EC 1.11.1.7) are implicated in a broad range of physiological processes throughout the plant life cycle. However, the unambiguous determination of the precise biological role of an individual class III peroxidase isoenzyme is still a difficult
Alok K Gupta et al.
BMC plant biology, 12, 158-158 (2012-09-04)
Silene latifolia and its pollinator, the noctuid moth Hadena bicruris, represent an open nursery pollination system wherein floral volatiles, especially veratrole (1, 2-dimethoxybenzene), lilac aldehydes, and phenylacetaldehyde are of key importance for floral signaling. Despite the important role of floral

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service