Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900766

Sigma-Aldrich

2,3-Pyridinedicarboxylic acid

Vetec, reagent grade, 98%

Synonym(s):

Quinolinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5NO4
CAS Number:
Molecular Weight:
167.12
Beilstein/REAXYS Number:
137110
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

188-190 °C (dec.) (lit.)

SMILES string

OC(=O)c1cccnc1C(O)=O

InChI

1S/C7H5NO4/c9-6(10)4-2-1-3-8-5(4)7(11)12/h1-3H,(H,9,10)(H,11,12)

InChI key

GJAWHXHKYYXBSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Moroni et al.
Journal of neurochemistry, 47(6), 1667-1671 (1986-12-01)
Quinolinic acid (QUIN), an excitotoxic tryptophan metabolite, has been identified and measured in human cerebrospinal fluid (CSF) using a mass-fragmentographic method. Furthermore, its content has been evaluated in frontal cortex obtained at autopsy from the cadavers of patients who died
Apsara Kandanearatchi et al.
The FEBS journal, 279(8), 1366-1374 (2012-01-21)
This brief review will first consider HIV associated neurocognitive disorder followed by the current understanding of its neuropathogenesis. Against this background the role of the kynurenine pathway will be detailed. Evidence both direct and indirect will be discussed for involvement
Valéry Combes et al.
Trends in parasitology, 28(8), 311-319 (2012-06-26)
Homeostasis implies constant operational defence mechanisms, against both external and internal threats. Infectious agents are prominent among such threats. During infection, the host elicits the release of a vast array of molecules and numerous cell-cell interactions are triggered. These pleiomorphic
Gilles J Guillemin et al.
Neuro-degenerative diseases, 2(3-4), 166-176 (2006-08-16)
The kynurenine pathway (KP) is a major route of L-tryptophan catabolism leading to production of several neurobiologically active molecules. Among them is the excitotoxin quinolinic acid (QUIN) that is known to be involved in the pathogenesis of several major inflammatory
Katherine R Leaver et al.
ACS chemical neuroscience, 3(2), 114-119 (2012-08-04)
There is evidence that excitotoxicity and prolonged microglial activation are involved in neuronal death in neurodegenerative disorders. Activated microglia express various molecules, including the translocator protein 18 kDa (TSPO; formerly known as the peripheral benzodiazepine receptor) on the outer mitochondrial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service