Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900761

Sigma-Aldrich

1-Indanone

Vetec, reagent grade, 98%

Synonym(s):

α-Hydrindone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein/REAXYS Number:
507957
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

bp

243-245 °C (lit.)

mp

38-40 °C (lit.)

density

1.103 g/mL at 25 °C (lit.)

SMILES string

O=C1CCc2ccccc12

InChI

1S/C9H8O/c10-9-6-5-7-3-1-2-4-8(7)9/h1-4H,5-6H2

Inchi Key

QNXSIUBBGPHDDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

233.0 °F - closed cup

flash_point_c

111.67 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service