Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900758

Sigma-Aldrich

Propionamide

Vetec, reagent grade, 97%

Synonym(s):

Propanamide, Propylamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CONH2
CAS Number:
Molecular Weight:
73.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

bp

213 °C (lit.)

mp

76-79 °C (lit.)

density

1.042 g/mL at 25 °C (lit.)

SMILES string

CCC(N)=O

InChI

1S/C3H7NO/c1-2-3(4)5/h2H2,1H3,(H2,4,5)

InChI key

QLNJFJADRCOGBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale W Sickles et al.
Toxicology and applied pharmacology, 222(1), 111-121 (2007-06-02)
The microtubule (MT) motor protein kinesin is a vital component of cells and organs expressing acrylamide (ACR) toxicity. As a mechanism of its potential carcinogenicity, we determined whether kinesins involved in cell division are inhibited by ACR similar to neuronal
Richard H Stadler
Advances in experimental medicine and biology, 561, 157-169 (2006-01-28)
This paper summarizes the progress made to date on acrylamide research pertaining to analytical methods, mechanisms of formation, and mitigation research in the major food categories. Initial difficulties with the establishment of reliable analytical methods have today in most cases
J D Stone et al.
Toxicology and applied pharmacology, 161(1), 50-58 (1999-11-24)
Neurofilament modification and accumulation, occurring in toxicant-induced neuropathies, has been proposed to compromise fast axonal transport and contribute to neurological symptoms or pathology. The current study compares the effects of the neurotoxicants acrylamide (ACR) and 2,5-hexanedione (2,5-HD) on the quantity
Ramesh Narayanan et al.
Molecular endocrinology (Baltimore, Md.), 22(11), 2448-2465 (2008-09-20)
Androgen receptor (AR) ligands are important for the development and function of several tissues and organs. However, the poor oral bioavailability, pharmacokinetic properties, and receptor cross-reactivity of testosterone, coupled with side effects, place limits on its clinical use. Selective AR
Mihail F Zilbermint et al.
Future oncology (London, England), 5(8), 1211-1220 (2009-10-27)
Cancer cachexia is a complex syndrome, affecting up to 60% of the approximately 1.4 million patients diagnosed with cancer each year in the USA. This condition is characterized by progressive deterioration of a patient's nutritional status, weight loss, anorexia, diminished

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service