Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900755

Sigma-Aldrich

Triethyl phosphate

Vetec, reagent grade, 98%

Synonym(s):

Ethyl phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3PO
CAS Number:
Molecular Weight:
182.15
Beilstein/REAXYS Number:
1705772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

6.28 (vs air)

vapor pressure

1 mmHg ( 40 °C)

product line

Vetec

assay

98%

autoignition temp.

845 °F

expl. lim.

10 %

refractive index

n20/D 1.403 (lit.)

bp

215 °C (lit.)

density

1.072 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(OCC)OCC

InChI

1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3

InChI key

DQWPFSLDHJDLRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

240.8 °F - closed cup

flash_point_c

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shujuan Meng et al.
Water research, 83, 248-257 (2015-07-15)
Alginate, a linear copolymer, is composed of 1,4-linked β-d-mannuronic acid (M) and α-l-guluronic acid (G), which are combined into homopolymeric blocks (MM-block and GG-block) and heteropolymeric block (MG-block). It has been widely used as a model foulant in various studies
T Salthammer et al.
Indoor air, 13(1), 49-52 (2003-03-01)
Organophosphate esters, halogenated and non-halogenated, are frequently used for fire protection of building materials. With regard to toxicological profiles it is desired to avoid human exposure in the indoor environment. Moreover, some hazardous volatile organic compounds detected in indoor air
B S Fales et al.
Journal of the American Society for Mass Spectrometry, 22(10), 1862-1871 (2011-09-29)
The gas-phase structures of protonated and sodium cationized complexes of triethyl phosphate, [TEP + H](+) and [TEP + Na](+), are examined via infrared multiple photon dissociation (IRMPD) action spectroscopy using tunable IR radiation generated by a free electron laser, a
Yuze Sun et al.
The Analyst, 134(7), 1386-1391 (2009-06-30)
We demonstrated rapid 2,4-dinitrotoluene (DNT) vapor detection at room temperature based on an optofluidic ring resonator (OFRR) sensor. With the unique on-column separation and detection features of OFRR vapor sensors, DNT can be identified from other interferences coexisting in the
Nele Van den Eede et al.
Environment international, 37(2), 454-461 (2010-12-24)
A new and efficient analytical method was developed and validated for the analysis of organophosphorus flame retardants (OPFRs) in indoor dust samples. This method involves an extraction step by ultrasonication and vortex, followed by extract clean-up with Florisil solid-phase extraction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service