Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900746

Sigma-Aldrich

p-Toluenesulfonamide

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2NH2
CAS Number:
Molecular Weight:
171.22
Beilstein/REAXYS Number:
472689
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

134-137 °C (lit.)

SMILES string

Cc1ccc(cc1)S(N)(=O)=O

InChI

1S/C7H9NO2S/c1-6-2-4-7(5-3-6)11(8,9)10/h2-5H,1H3,(H2,8,9,10)

InChI key

LMYRWZFENFIFIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

395.6 °F - closed cup

flash_point_c

202 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dingben Chen et al.
The Journal of organic chemistry, 75(16), 5768-5771 (2010-07-30)
A copper-catalyzed cascade method has been developed to synthesize the 2H-benzo[b][1,4]thiazin-3(4H)-ones from 2-halo-N-(2-halophenyl)-acetamides 1 and AcSH via the S(N)2/deacetylation/coupling process, and to synthesize the quinoxalin-2(1H)-ones from 1 and TsNH(2) via the S(N)2/coupling/desulfonation process. The target products were obtained with diversity
Yang Yang et al.
Organic letters, 13(20), 5608-5611 (2011-09-16)
BF(3)·OEt(2)-catalyzed direct cyanation of indoles and pyrroles using a less toxic, bench-stable, and easily handled electrophilic cyanating agent N-cyano-N-phenyl-para-toluenesulfonamide (NCTS) affords 3-cyanoindoles and 2-cyanopyrroles in good yields with excellent regioselectivity. The substrate scope is broad with respect to indoles and
Christian Lorber et al.
Inorganic chemistry, 46(8), 3192-3202 (2007-03-17)
p-Toluenesulfonamide (p-TsNH2) was successfully employed as an imido ligand precursor in the synthesis of highly air- and moisture-sensitive titanium(IV) and vanadium(IV) complexes. Reaction of M(NMe2)4 (M = Ti, V) with TsNH2 in toluene afforded [M(micro-NTs)(NMe2)2]2 dimer complexes (M = Ti
J R Meinertz et al.
Journal of AOAC International, 84(5), 1332-1336 (2001-10-17)
Chloramine-T is an effective drug for controlling fish mortality caused by bacterial gill disease. As part of the data required for approval of chloramine-T use in aquaculture, depletion of the chloramine-T marker residue (para-toluenesulfonamide; p-TSA) from edible fillet tissue of
J R Meinertz et al.
Journal of AOAC International, 82(5), 1064-1070 (1999-10-08)
Chloramine-T (N-sodium-N-chloro-p-toluene-sulfonamide) is a candidate therapeutic drug for treating bacterial gill disease, a predominant disease of a variety of fish species. Research has been initiated to obtain the U.S. Food and Drug Administration's (FDA) approval for the use of chloramine-T

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service