Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900744

Sigma-Aldrich

2-Adamantanone

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
Beilstein/REAXYS Number:
1210235
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

256-258 °C (subl.) (lit.)

SMILES string

O=C1C2CC3CC(C2)CC1C3

InChI

1S/C10H14O/c11-10-8-2-6-1-7(4-8)5-9(10)3-6/h6-9H,1-5H2/t6-,7+,8-,9+

InChI key

IYKFYARMMIESOX-SPJNRGJMSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassen Jaafar et al.
Dalton transactions (Cambridge, England : 2003), 40(1), 92-106 (2010-11-19)
We report that the oxygen sensitivity of some Fe(II) complexes with tripodal ligands can be used, with benefit, in the oxidation of cyclohexane under mild conditions. Depending on the solvent, two very different reaction pathways are involved, which share the
Yulan Chen et al.
Nature chemistry, 4(7), 559-562 (2012-06-22)
Nature uses mechanochemical transduction processes to achieve diverse and vital functions, such as hearing, cellular adhesion and gating of ion channels. One fascinating example of biological mechanotransduction is the emission of light on mechanical stimulation. However, molecular-level transduction of force
Roman Davydov et al.
Journal of the American Chemical Society, 127(5), 1403-1413 (2005-02-03)
EPR/ENDOR studies have been carried out on oxyferrous cytochrome P450cam one-electron cryoreduced by gamma-irradiation at 77 K in the absence of substrate and in the presence of a variety of substrates including its native hydroxylation substrate, camphor (a), and the
Solid-state and solution studies of a tetrameric capsule and its guests.
Darren W Johnson et al.
Angewandte Chemie (International ed. in English), 41(20), 3793-3796 (2002-10-19)
E Mariani et al.
Il Farmaco; edizione scientifica, 35(5), 430-440 (1980-05-01)
The synthesis of (+/-)-1-(adamantan-2-yl)-2-propanamine (VI), its N,N-dimethyl derivative (VII) and other amines (VIII), (IX) and (X), starting from 2-adamantanone, is described. Some of these amines showed antiparkinson activity in mice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service