Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900743

Sigma-Aldrich

γ-Caprolactone

Vetec, reagent grade, 98%

Synonym(s):

γ-Ethyl-γ-butyrolactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein/REAXYS Number:
6114822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.439 (lit.)

bp

219 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

CCC1CCC(=O)O1

InChI

1S/C6H10O2/c1-2-5-3-4-6(7)8-5/h5H,2-4H2,1H3

InChI key

JBFHTYHTHYHCDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olga Khersonsky et al.
Biochemistry, 44(16), 6371-6382 (2005-04-20)
PON1 is the best-studied member of a family of enzymes called serum paraoxonases, or PONs, identified in mammals (including humans) and other vertebrates as well as in invertebrates. PONs exhibit a range of important activities, including drug metabolism and detoxification
H Thoma et al.
Journal of chromatography, 309(1), 17-32 (1984-07-13)
A two-step ultrafiltration method combined with anion-exchange chromatography is described for the separation of lipophilic and strongly lipophobic acids occurring in human blood, serum and plasma. After treatment with diazomethane, the acid fractions are separated further by gas chromatography. The
Profile of volatile metabolites in human urine.
A Zlatkis et al.
Clinical chemistry, 17(7), 592-594 (1971-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service