Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900739

Sigma-Aldrich

o-Tolylacetic acid

Vetec, reagent grade, 98%

Synonym(s):

2-Methylphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4CH2CO2H
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

88-90 °C (lit.)

SMILES string

Cc1ccccc1CC(O)=O

InChI

1S/C9H10O2/c1-7-4-2-3-5-8(7)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

RZWGTXHSYZGXKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yohei Okubo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(48), 15837-15846 (2015-12-04)
The endoplasmic reticulum (ER) plays crucial roles in intracellular Ca(2+) signaling, serving as both a source and sink of Ca(2+), and regulating a variety of physiological and pathophysiological events in neurons in the brain. However, spatiotemporal Ca(2+) dynamics within the
Ai-Hui Tang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(9), 3938-3945 (2015-03-06)
The Fmr1 knock-out mouse model of fragile X syndrome (Fmr1(-/y)) has an epileptogenic phenotype that is triggered by group I metabotropic glutamate receptor (mGluR) activation. We found that a membrane-permeable peptide that disrupts mGluR5 interactions with long-form Homers enhanced mGluR-induced
Samuel W Cramer et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(14), 5664-5679 (2015-04-10)
The Ca(2+) channelopathies caused by mutations of the CACNA1A gene that encodes the pore-forming subunit of the human Cav2.1 (P/Q-type) voltage-gated Ca(2+) channel include episodic ataxia type 2 (EA2). Although, in EA2 the emphasis has been on cerebellar dysfunction, patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service