Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900738

Sigma-Aldrich

N-Phenylurea

Vetec, reagent grade, 97%

Synonym(s):

Phenylcarbamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCONH2
CAS Number:
Molecular Weight:
136.15
Beilstein/REAXYS Number:
1934615
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

vapor density

>1 (vs air)

product line

Vetec

assay

97%

mp

145-147 °C (lit.)

SMILES string

NC(=O)Nc1ccccc1

InChI

1S/C7H8N2O/c8-7(10)9-6-4-2-1-3-5-6/h1-5H,(H3,8,9,10)

InChI key

LUBJCRLGQSPQNN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Palma et al.
The Science of the total environment, 488-489, 208-219 (2014-05-20)
The purpose of this study was to evaluate the potential impact of the pesticides detected in the Alqueva reservoir (Guadiana Basin, South Iberian Peninsula) on the aquatic organisms belonging to this ecosystem. For this purpose, the occurrence and risk assessment
Pankaj Aggarwal et al.
Journal of chromatography. A, 1364, 96-106 (2014-09-07)
Highly cross-linked monolithic networks (i.e., polyethylene glycol diacrylate, PEGDA) synthesized from monomers containing varying ethylene oxide chain lengths were fabricated inside fused silica capillary columns for use in liquid chromatography (LC) of small molecules. Tergitol was used as a surfactant
Wei-Hsiang Chen et al.
Environmental science. Processes & impacts, 17(12), 2092-2100 (2015-11-06)
The objective of this study was to investigate the formation of different nitrosamines during chlorination or chloramination (chlor(am)ination) of five phenylurea herbicides (fluometuron, diuron, linuron, metobromuron, and propanil), with the effects of disinfection approaches, additional inorganic nitrogen, and reaction pH
Shu-Ling Lin et al.
Talanta, 147, 199-206 (2015-11-26)
In this study, recently developed 1,6-hexanediol ethoxylate diacrylate (HEDA)-based polymeric monoliths were utilized as sorbents for efficient extraction of phenylurea herbicides (PUHs) from water samples. The HEDA-based monolithic sorbents were prepared in a fused silica capillary (0.7mm i.d., 4.5-cm long)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service