Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900737

Sigma-Aldrich

4-Mercaptophenol

Vetec, reagent grade, 89%

Synonym(s):

4-Hydroxythiophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4OH
CAS Number:
Molecular Weight:
126.18
Beilstein/REAXYS Number:
2039306
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

89%

bp

149-150 °C/25 mmHg (lit.)

mp

33-35 °C (lit.)

SMILES string

Oc1ccc(S)cc1

InChI

1S/C6H6OS/c7-5-1-3-6(8)4-2-5/h1-4,7-8H

InChI key

BXAVKNRWVKUTLY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabel Freitag et al.
Journal of biomedical optics, 20(5), 55002-55002 (2015-05-06)
Identification of tumor and normal cells is a promising application of Raman spectroscopy. The throughput of Raman-assisted cell sorting is limited by low sensitivity. Surface-enhanced Raman spectroscopy (SERS) is a well-recognized candidate to increase the intensity of Raman signals of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service