Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900734

Sigma-Aldrich

Pyridine hydrochloride

Vetec, reagent grade, 98%

Synonym(s):

Pyridinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N · HCl
CAS Number:
Molecular Weight:
115.56
Beilstein/REAXYS Number:
3615340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

222-224 °C (lit.)

mp

145-147 °C (lit.)

SMILES string

Cl[H].c1ccncc1

InChI

1S/C5H5N.ClH/c1-2-4-6-5-3-1;/h1-5H;1H

InChI key

AOJFQRQNPXYVLM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatyana N Sevastyanova et al.
Molecular pharmacology, 86(5), 492-504 (2014-08-13)
Metabotropic glutamate receptors (mGluRs) function as dimers. Recent work suggests that mGluR1 and mGluR5 may physically interact, but the nature and functional consequences of this relationship have not been addressed. In this study, the functional and pharmacological consequences of this
Wen Zhou et al.
Inorganic chemistry, 52(5), 2271-2273 (2013-02-20)
The reaction of 2,2'-bipyridine (bpy) with monomeric chromium(II) precursors was used to prepare the S = 1 complexes Cr(tBu-acac)2(bpy) (1) and (η(5)-Cp)(η(1)-Cp)Cr(bpy) (3), as well as the S = 2 compound Cr[N(SiMe3)2]2(bpy) (4). The crystallographically determined bond lengths indicate that
Vandana Bhalla et al.
Dalton transactions (Cambridge, England : 2003), 42(13), 4450-4455 (2013-01-26)
Nanoaggregates of a supramolecular ensemble of triphenylene derivative 3 and Cu(2+) ions exhibit 'turn-on' response towards CN(-) ions in an aqueous medium and can detect the trace amount of inorganic cyanide ions (NaCN) in tap water and blood serum milieu.
Synthetic studies on pseudo-dimeric Lycopodium alkaloids: total synthesis of complanadine B.
James N Newton et al.
Angewandte Chemie (International ed. in English), 52(6), 1726-1730 (2013-01-12)
Nathalie Ségaud et al.
Inorganic chemistry, 52(2), 691-700 (2013-01-11)
We report the synthesis, characterization, and solution chemistry of a series of new Fe(II) complexes based on the tetradentate ligand N-methyl-N,N'-bis(2-pyridyl-methyl)-1,2-diaminoethane or the pentadentate ones N,N',N'-tris(2-pyridyl-methyl)-1,2-diaminoethane and N,N',N'-tris(2-pyridyl-methyl)-1,3-diaminopropane, modified by propynyl or methoxyphenyltriazolyl groups on the amino functions. Six of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service