Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900730

Sigma-Aldrich

Phenacetin

Vetec, reagent grade, 98%

Synonym(s):

1-Acetyl-p-phenetidin, 4′-Ethoxyacetanilide, N-(4-Ethoxyphenyl)acetamide, p-Acetophenetidide, Acetophenetidin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OC2H5
CAS Number:
Molecular Weight:
179.22
Beilstein/REAXYS Number:
1869238
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

133-136 °C (lit.)

SMILES string

CCOc1ccc(NC(C)=O)cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)

InChI key

CPJSUEIXXCENMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Substrate of CYP1A2 and CYP2D6.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Kobayashi et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(4), 671-679 (2011-12-31)
Human arylacetamide deacetylase (AADAC) is a major esterase responsible for the hydrolysis of clinical drugs such as flutamide, phenacetin, and rifampicin. Thus, AADAC is considered to be a relevant enzyme in preclinical drug development, but there is little information about
Xin Wang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(3-4), 348-354 (2011-11-08)
Danshen (Salvia miltiorrhiza Bunge) as a famous Traditional Chinese medicine is widely used in the treatment of cardiovascular and cerebrovascular diseases in the world. Danshen tincture (DT), extracted from Danshen root with a mixture of water and alcohol, is a
Heloisa N Bordallo et al.
Molecular pharmaceutics, 9(9), 2434-2441 (2012-07-25)
This study centers on the use of inelastic neutron scattering as an alternative tool for physical characterization of solid pharmaceutical drugs. On the basis of such approach, relaxation processes in the pharmaceutical compound phenacetin (p-ethoxyacetanilide, C(10)H(13)NO(2)) were evidenced on heating
Yuki Kobayashi et al.
Biochemical pharmacology, 84(9), 1196-1206 (2012-09-04)
Phenacetin has been used as an analgesic antipyretic but has now been withdrawn from the market due to adverse effects such as methemoglobinemia and renal failure. It has been suggested that metabolic activation causes these adverse effects; yet, the precise
Kazuo Takayama et al.
Journal of hepatology, 57(3), 628-636 (2012-06-05)
Hepatocyte-like cells differentiated from human embryonic stem cells (hESCs) and induced pluripotent stem cells (hiPSCs) can be utilized as a tool for screening for hepatotoxicity in the early phase of pharmaceutical development. We have recently reported that hepatic differentiation is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service