Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900728

Sigma-Aldrich

Terephthalonitrile

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CN)2
CAS Number:
Molecular Weight:
128.13
Beilstein/REAXYS Number:
1072210
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

221-225 °C (lit.)

SMILES string

N#Cc1ccc(cc1)C#N

InChI

1S/C8H4N2/c9-5-7-1-2-8(6-10)4-3-7/h1-4H

InChI key

BHXFKXOIODIUJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suchandra Chatterjee et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 60(1-2), 97-102 (2003-12-13)
With the advent of spin chemistry, magnetic field effect (MFE) on exciplex luminescence has emerged as an important domain of research. MFE is a diffusion controlled phenomenon and hence is solvent dielectric (epsilon) dependent. It maximizes at a particular epsilon
T Sengupta et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 57A(5), 1103-1110 (2001-05-26)
The exciplex between all-s-trans-1,4-diphenylbuta-1,3-diene and 1,4-dicyanobenzene has been studied by steady state fluorescence along with the magnetic field effect (MFE) and compared with the other alpha,omega-diphenyl polyenes. The exciplex formation and magnetic field effect are dictated by the chain length
Christian Ochsenfeld et al.
Solid state nuclear magnetic resonance, 22(2-3), 128-153 (2002-12-10)
A study of a host-guest system consisting of a naphthalene-spaced tweezer with a 1,4 dicyanobenzene guest molecule is presented. The complex is investigated using a combination of quantum-chemical calculations and solid-state NMR experiments. The advantages of such an approach are
Liheng Feng et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(3), 646-650 (2007-03-31)
A novel luminescent metal complex, (MQPF)3Al2, with 8-hydroxyquinoline aluminum and 9,9-diphenylfluorene was synthesized. The optical properties were investigated by UV-vis absorption and fluorescence emission spectra. The results showed that the luminescence quantum yield of (MQPF)3Al2 was 0.612 in THF and
Felix Eberle et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 11(13), 2951-2956 (2010-08-18)
Self-assembled monolayers of 1,4-dicyanobenzene on Au(111) electrodes are studied by cyclic voltammetry, in-situ STM and ex-situ XPS. High-resolution STM images reveal a long-range order of propeller-like assemblies each of which consists of three molecules, all lying flat on the gold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service