Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900726

Sigma-Aldrich

4-(2-Hydroxyethyl)morpholine

Vetec, reagent grade, 98%

Synonym(s):

2-Morpholinoethanol, 4-Morpholineethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H13NO2
CAS Number:
Molecular Weight:
131.17
Beilstein/REAXYS Number:
104375
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

refractive index

n20/D 1.476 (lit.)

bp

227 °C/757 mmHg (lit.)

density

1.083 g/mL at 25 °C (lit.)

SMILES string

OCCN1CCOCC1

InChI

1S/C6H13NO2/c8-4-1-7-2-5-9-6-3-7/h8H,1-6H2

InChI key

KKFDCBRMNNSAAW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

208.4 °F - closed cup

flash_point_c

98 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoine Frère et al.
Biomacromolecules, 16(3), 769-779 (2015-01-21)
RNAi therapeutics are promising therapeutic tools that have sparked the interest of many researchers. In an effort to provide a safe alternative to PEI, we have designed a series of new guanidinium- and morpholino-functionalized biocompatible and biodegradable polycarbonate vectors. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service