Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900725

Sigma-Aldrich

4-Nitrobenzoic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CO2H
CAS Number:
Molecular Weight:
167.12
Beilstein/REAXYS Number:
973593
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

237-240 °C (lit.)

SMILES string

OC(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H5NO4/c9-7(10)5-1-3-6(4-2-5)8(11)12/h1-4H,(H,9,10)

InChI key

OTLNPYWUJOZPPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

458.6 °F - closed cup

flash_point_c

237 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhifen Lin et al.
Chemical research in toxicology, 16(10), 1365-1371 (2003-10-21)
Quantitative structure-activity relationship (QSAR) approaches are proposed in this study to predict the joint effects of mixture toxicity. The initial investigation studies the joint effects between cyanogenic toxicants and aldehydes to Photobacterium phosphoreum. Joint effects are found to result from
Ulrika Forsman et al.
Nature chemical biology, 6(7), 515-517 (2010-06-08)
Coenzyme Q (Q) is an electron transporter in the respiratory chain and a lipid-soluble antioxidant that decreases in humans with age. Here we show that 4-nitrobenzoate inhibited 4-hydroxybenzoate:polyprenyl transferase (Coq2) in a competitive manner and dose-dependently decreased Q in mammalian
Lisheng Zhang et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 363-366 (2012-04-13)
Well-ordered silver nano arrays, prepared using anodic aluminum oxide (AAO) template by laser molecular beam epitaxy (L-MBE) method, were adhered to the work electrode using conductive adhesive to be a Surface-enhanced Raman scattering (SERS) substrate. Variable SERS signals of paranitrobenzoic
Li Lin et al.
Chemosphere, 78(1), 66-71 (2009-10-23)
This paper describes the microwave (MW) treatment of soil contaminated by chloramphenicol (CAP), using granular activated carbon (GAC) as MW absorbent. It was found that the addition of GAC effectively increased the temperature of soil. Large MW power and GAC
J A Gómez-Vidal et al.
Organic letters, 3(16), 2477-2479 (2001-08-03)
[reaction: see text] A mild and selective cleavage of p-nitrobenzoic esters by sodium azide in methanol is reported. This new methodology is mild enough for use with acid- or base-sensitive compounds. No elimination byproducts are formed. Fmoc- and trifluoroacetyl-amino protecting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service