Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900718

Sigma-Aldrich

2-Methylbenzimidazole

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2
CAS Number:
Molecular Weight:
132.16
Beilstein/REAXYS Number:
112264
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

175-177 °C (lit.)

SMILES string

Cc1nc2ccccc2[nH]1

InChI

1S/C8H8N2/c1-6-9-7-4-2-3-5-8(7)10-6/h2-5H,1H3,(H,9,10)

InChI key

LDZYRENCLPUXAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Sarcina et al.
FEMS microbiology letters, 191(1), 25-29 (2000-09-27)
The bacterial cell division protein FtsZ is a structural analogue of tubulin. Bacterial mutants in which the ftsZ gene is inactivated are unable to divide. Numerous inhibitors of tubulin assembly are known, some of which are used as fungicides. The
M Tahir Güllüoglu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(2), 107-114 (2010-04-17)
The FT-IR and FT-Raman spectra of 2-methylbenzimidazole (2-MB) and 5-methylbenzimidazole (5-MB) molecules have been recorded between 400-4000 cm(-1) and 50-3500 cm(-1) region, respectively. The molecular geometry and vibrational frequencies of 2- and 5-MB molecules in the ground state have been
A E Renauld et al.
Comparative biochemistry and physiology. Part C, Pharmacology, toxicology & endocrinology, 122(2), 273-281 (1999-04-06)
Seven modulators of mammalian monooxygenase activity were screened for their ability to selectively stimulate or inhibit in vitro monooxygenase activities of hepatic microsomes from mallard ducklings treated with phenobarbital, beta-naphthoflavone, 3,3',4,4',5-pentachlorobiphenyl or vehicle. Microsomes were assayed fluorometrically for four monooxygenases:
A Svoboda et al.
Canadian journal of microbiology, 43(10), 945-953 (1997-12-13)
Both living and fixed cells of Schizosaccharomyces japonicus var. versatilis showed thread-like mitochondria when studied by phase-contrast and fluorescence microscopy. In the interphase cells, mitochondria extended from pole to pole and converged towards the growing tips. The mitochondrial threads did
Ahmed O H El-Nezhawy et al.
Bioorganic & medicinal chemistry, 21(7), 1661-1670 (2013-03-05)
A new series of novel benzimidazole derivatives containing substituted pyrid-2-yl moiety and polyhydroxy sugar conjugated to the N-benzimidazole moiety has been synthesized and evaluated as orally bioavailable anti-inflammatory agents with anti-ulcerogenic activity. The anti-inflammatory and anti-ulcerogenic activities of these compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service