V900717
2-Indanone
Vetec™, reagent grade, 98%
Synonym(s):
β-Hydrindone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H8O
CAS Number:
Molecular Weight:
132.16
Beilstein/REAXYS Number:
636550
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
reagent grade
product line
Vetec™
assay
98%
mp
51-54 °C (lit.)
storage temp.
2-8°C
SMILES string
O=C1Cc2ccccc2C1
InChI
1S/C9H8O/c10-9-5-7-3-1-2-4-8(7)6-9/h1-4H,5-6H2
InChI key
UMJJFEIKYGFCAT-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Vetec is a trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
212.0 °F - closed cup
flash_point_c
100 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service