Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900716

Sigma-Aldrich

2-Hydroxyphenylacetic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
908000
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

145-147 °C (lit.)

SMILES string

OC(=O)Cc1ccccc1O

InChI

1S/C8H8O3/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4,9H,5H2,(H,10,11)

InChI key

CCVYRRGZDBSHFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Y Hsu et al.
Journal of pharmaceutical and biomedical analysis, 132, 109-116 (2016-10-05)
A high performance liquid chromatography-diode array detection-tandem mass spectrometry method (HPLC-DAD-MS/MS) was developed for simultaneous determination of phenolic acids and flavonoids in djulis (Chenopodium formosanum Koidz.), a traditional Chinese herb reported to possess vital biological activities. A high yield of
M Bayat-Sarmadi et al.
Molecular and cellular endocrinology, 92(1), 127-134 (1993-03-01)
Prolactin has many known functions and one of them is to induce the expression of milk protein gene expression in the mammary gland. Specific membrane receptors have been recently characterized but the transduction mechanism involved in the transfer of the
Jeffrey D Vassallo et al.
Toxicological sciences : an official journal of the Society of Toxicology, 80(2), 249-257 (2004-05-14)
Hepatotoxicity of coumarin is attributed to metabolic activation to an epoxide intermediate, coumarin 3,4-epoxide (CE). However, whereas rats are most susceptible to coumarin-induced hepatotoxicity, formation of CE is greatest in mouse liver microsomes, a species showing little evidence of hepatotoxicity.
S L Born et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 41(2), 247-258 (2002-12-14)
Coumarin, a well recognized rat hepatotoxicant, also causes acute, selective necrosis of terminal bronchiolar Clara cells in the mouse lung. Further, chronic oral gavage administration of coumarin at 200 mg/kg, a dose that causes Clara cell death, resulted in a
D Ratanasavanh et al.
Fundamental & clinical pharmacology, 10(6), 504-510 (1996-01-01)
We compared the cytotoxic effect of coumarin and its derivatives, 7-hydroxycoumarin (7-OHC), 4-hydroxycoumarin (4-OHC), o-hydroxyphenyl acetic acid (OHPAA) and o-coumaric acid (CA), on cultured hepatocytes from human, rat, mouse and rabbit liver. At 10(-5) and 5 x 10(-5) M, coumarin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service