Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900710

Supelco

Antipyrine

Vetec, reagent grade, 98%

Synonym(s):

2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Beilstein/REAXYS Number:
157775
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

109-111 °C (lit.)

SMILES string

CN1N(C(=O)C=C1C)c2ccccc2

InChI

1S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3

Inchi Key

VEQOALNAAJBPNY-UHFFFAOYSA-N

Gene Information

human ... PTGS1(5742)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Line Mathiesen et al.
PloS one, 15(6), e0233979-e0233979 (2020-06-04)
Exposure to maternal stress during pregnancy can have adverse effects on the fetus, which has potential long-term effects on offspring´s development and health. We investigated the kinetics and metabolism of the hormones and amino acids: cortisol, cortisone, tryptophan and serotonin
Gro Klitgaard Povlsen et al.
BMC neuroscience, 14, 34-34 (2013-03-19)
Upregulation of vasoconstrictor receptors in cerebral arteries, including endothelin B (ETB) and 5-hydroxytryptamine 1B (5-HT(1B)) receptors, has been suggested to contribute to delayed cerebral ischemia, a feared complication after subarachnoid hemorrhage (SAH). This receptor upregulation has been shown to be
Joy Eliaerts et al.
Drug testing and analysis, 9(10), 1480-1489 (2016-12-16)
Traditionally, fast screening for the presence of cocaine in unknown powders is performed by means of colour tests. The major drawbacks of these tests are subjective colour evaluation depending on the operator ('50 shades of blue') and a lack of
Factors influencing antipyrine elimination.
I H Stevenson
British journal of clinical pharmacology, 4(3), 261-265 (1977-06-01)
Antipyrine as a model drug to study hepatic drug-metabolizing capacity.
H E Poulsen et al.
Journal of hepatology, 6(3), 374-382 (1988-06-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service