Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900706

Sigma-Aldrich

DL-Methionine

98%, Vetec

Synonym(s):

(±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Beilstein/REAXYS Number:
636185
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

Product Name

DL-Methionine, Vetec, reagent grade, 98%

grade

reagent grade

product line

Vetec

assay

98%

form

powder

color

white

mp

280 °C (dec.) (lit.)

SMILES string

CSCCC(N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shawn Flanagan et al.
Antimicrobial agents and chemotherapy, 59(1), 178-185 (2014-10-22)
Prolonged treatment with the oxazolidinone linezolid is associated with myelosuppression, lactic acidosis, and neuropathies, toxicities likely caused by impairment of mitochondrial protein synthesis (MPS). To evaluate the potential of the novel oxazolidinone tedizolid to cause similar side effects, nonclinical and
Camilla Schalin-Jäntti et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(5), 739-747 (2013-04-05)
All patients with primary hyperparathyroidism should undergo localization studies before reoperation, but it is not known which method is most accurate. The purpose of this prospective study was to compare the performance of planar scintigraphy with (123)I/(99m)Tc-sestamibi, (99m)Tc-sestamibi SPECT (SPECT/CT)
Gavin C Preston et al.
The EMBO journal, 34(15), 2008-2024 (2015-07-03)
Myc controls the metabolic reprogramming that supports effector T cell differentiation. The expression of Myc is regulated by the T cell antigen receptor (TCR) and pro-inflammatory cytokines such as interleukin-2 (IL-2). We now show that the TCR is a digital
Leen Mathys et al.
PloS one, 9(6), e101181-e101181 (2014-06-27)
N-linked glycans covering the surface of the HIV-1 glycoprotein gp120 are of major importance for the correct folding of this glycoprotein. Of the, on average, 24 N-linked glycans present on gp120, the glycan at Asn260 was reported to be essential
Mario Ost et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(4), 1314-1328 (2014-12-11)
Recent studies on mouse and human skeletal muscle (SM) demonstrated the important link between mitochondrial function and the cellular metabolic adaptation. To identify key compensatory molecular mechanisms in response to chronic mitochondrial distress, we analyzed mice with ectopic SM respiratory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service