Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900704

Sigma-Aldrich

Methylamine hydrochloride

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NH2 · HCl
CAS Number:
Molecular Weight:
67.52
Beilstein/REAXYS Number:
3588822
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

225-230 °C/15 mmHg (lit.)

mp

231-233 °C (lit.)

SMILES string

Cl[H].CN

InChI

1S/CH5N.ClH/c1-2;/h2H2,1H3;1H

InChI key

NQMRYBIKMRVZLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas S Hofer et al.
Molecular bioSystems, 8(11), 2891-2900 (2012-08-01)
Molecular dynamics simulations have been performed to investigate the binding of tris(hydroxymethyl)-aminomethane to the surface of the core domain of the mouse cellular tumor antigen p53 employing the GROMOS and 53A6 force field parameter sets. A close investigation of the
Michaela Frybortova et al.
Journal of inorganic biochemistry, 126, 46-54 (2013-06-19)
In order to shed light on the mechanism that underlies activity of bifunctional mononuclear Pt(II) analogs of transplatin we examined in the present work a DNA binding mode of the analog of transplatin, namely trans-[Pt(CH3NH2)2Cl2], in which NH3 groups were
Marco Dionisio et al.
Journal of the American Chemical Society, 134(15), 6540-6543 (2012-04-06)
Single-walled carbon nanotubes (SWCNTs) have been functionalized with highly selective tetraphosphonate cavitand receptors. The binding of charged N-methylammonium species to the functionalized SWCNTs was analyzed by X-ray photoelectron spectroscopy and confirmed by (31)P MAS NMR spectroscopy. The cavitand-functionalized SWCNTs were
Elisabeth Schwaiger et al.
Transplantation, 97(12), 1279-1285 (2014-03-14)
Luminex-based anti-HLA IgG detection on single-antigen flow beads (SAFB) represents a valuable tool for characterization of allosensitization patterns. Assay interpretation, however, may be impeded by false-low test results caused by the prozone effect. Recent experimental data have related this artifact
Jing-Jing Fang et al.
Waste management (New York, N.Y.), 32(7), 1401-1410 (2012-04-07)
This study investigated the odor compounds from different areas in a landfill site, which included the municipal solid waste (MSW)-related area, the leachate-related area and the sludge-related area. Nine sampling points were placed and 35 types of odorous substances were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service