Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900702

Sigma-Aldrich

3-Methoxybenzoic acid

Vetec, reagent grade, 98%

Synonym(s):

m-Anisic acid, m-Methylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
508838
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

170-172 °C/10 mmHg (lit.)

mp

105-107 °C (lit.)

SMILES string

COc1cccc(c1)C(O)=O

InChI

1S/C8H8O3/c1-11-7-4-2-3-6(5-7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

XHQZJYCNDZAGLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thi-Huu Nguyen et al.
Organic letters, 7(12), 2445-2448 (2005-06-04)
[reaction: see text] If employed in THF at 0 degrees C, LTMP metalates meta-anisic acid at the doubly activated position. In contrast, n-BuLi/t-BuOK deprotonates position C-4 preferentially at low temperature. Functionalization at C-6 requires protection of the C-2 site beforehand.
N Dodoff et al.
Journal of inorganic biochemistry, 54(3), 221-233 (1994-05-15)
The complexes [Pt(bah)2X2], [Pt(NH3)(bah)Cl2].0.5H2O, [Pt(mbah)2X2], and [Pt(NH3)(mbah)Cl2] (bah = benzoic acid hydrazide, mbah = 3-methoxybenzoic acid hydrazide; X = Cl, Br, I) have been prepared and characterized by elemental analysis, electric conductivity, IR, 1H NMR, and electronic spectra. A cis-square
K A DeWeerd et al.
Applied and environmental microbiology, 54(5), 1237-1242 (1988-05-01)
O-methyl substituents of aromatic compounds can provide C1 growth substrates for facultative and strict anaerobic bacteria isolated from diverse environments. The mechanism of the bioconversion of methoxylated benzoic acids to the hydroxylated derivatives was investigated with a model substrate and
D H Lee et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(9), 909-916 (1999-11-05)
1. 2-(Allylthio)pyrazine (2-AP) has been demonstrated to protect the liver against toxicants by inhibiting CYP2E1 activity. Since 2-mercaptopyrazine (2-MP) is presumed to be a metabolite of 2-AP, the experiments were performed to determine whether rat liver microsomal and/or cytosolic preparations
P E Devaraj et al.
Molecular and cellular biochemistry, 124(2), 175-181 (1993-07-21)
In mouse fibroblasts stimulated from quiescence into proliferation by serum the induction of expression of the c-myc proto-oncogene is strongly stimulated by 3-methoxybenzamide. Similar superinduction effects are seen with related compounds such as 3-aminobenzamide and the acid analogues, 3-anisic acid

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service