Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900700

Sigma-Aldrich

2-Methoxybenzoic acid

Vetec, reagent grade, 98%

Synonym(s):

O-Methylsalicylic acid, o-Anisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
509929
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

98-100 °C (lit.)

SMILES string

COc1ccccc1C(O)=O

InChI

1S/C8H8O3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3,(H,9,10)

InChI key

ILUJQPXNXACGAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

291.2 °F - closed cup

flash_point_c

144 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Sasaki et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 50(5), 905-909 (1999-04-24)
For in vivo measurement of the hydroxyl radical (.OH), we synthesized [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid by carboxylation of 2-bromomagnesiumanisol using [11C]CO2. The radiochemical yield of [11C]salicylic acid, [11C]O-acetylsalicylic acid and [11C]2-methoxybenzoic acid calculated from trapped [11C]CO2 in
Quan Zhou et al.
The Journal of organic chemistry, 73(20), 8049-8056 (2008-09-20)
Mander reductive alkylation of methyl 2-methoxybenzoate with prenyl bromide and hydrolysis of the enol ether afforded methyl 6-oxo-1-prenyl-2-cyclohexenecarboxylate. This was converted in five steps (reduction of the ketone, saponification, iodolactonization, ozonolysis, and intramolecular aldol reaction) to a spiro lactone cyclopentenal.
M Gil et al.
Biodegradation, 11(1), 49-53 (2001-02-24)
Aromatic carboxylic acids substituted with methoxylated groups are among the most abundant products in "alpechin", the wastes resulting from pressing olives to obtain olive oil. Degradation of o-methoxybenzoate by an stable consortium made of a gram positive bacterium, Arthrobacter oxydans
Christoph Böttcher et al.
Analytical chemistry, 79(4), 1507-1513 (2007-02-15)
The coupling of liquid chromatography to electrospray ionization quadrupole time-of-flight mass spectrometry can be a powerful tool for metabolomics, i.e., the comprehensive detection of low molecular weight compounds in biological systems. There have, however, been doubts about the feasibility and
B Uma et al.
Toxicon : official journal of the International Society on Toxinology, 38(8), 1129-1147 (2000-03-10)
The basic phospholipase A(2) (VRV-PL-VIIIa) from Vipera russelli venom induces multiple toxic effects including neurotoxicity, myotoxicity, edema and hemorrhage. This phospholipase A(2) has been extensively characterized for its pharmacological properties except for hemorrhagic activity. In the present investigation, the lung

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service