Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900697

Sigma-Aldrich

Benzyltriethylammonium chloride

Vetec, reagent grade, 98%

Synonym(s):

N,N,N-Triethyl-N-benzylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2N(Cl)(C2H5)3
CAS Number:
Molecular Weight:
227.77
Beilstein/REAXYS Number:
3574984
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

190-192 °C (dec.) (lit.)

SMILES string

[Cl-].CC[N+](CC)(CC)Cc1ccccc1

InChI

1S/C13H22N.ClH/c1-4-14(5-2,6-3)12-13-10-8-7-9-11-13;/h7-11H,4-6,12H2,1-3H3;1H/q+1;/p-1

InChI key

HTZCNXWZYVXIMZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

>527.0 °F - closed cup

flash_point_c

> 275 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Petr Lazar et al.
Small (Weinheim an der Bergstrasse, Germany), 11(31), 3790-3796 (2015-05-06)
Halogen functionalization of graphene is an important branch of graphene research as it provides opportunities to tailor the band gap and catalytic properties of graphene. Monovalent C-X bond obviates pitfalls of functionalization with atoms of groups 13, 15, and 16
Dennis Åsberg et al.
Journal of chromatography. A, 1401, 52-59 (2015-05-25)
The importance of the generated temperature and pressure gradients in ultra-high-pressure liquid chromatography (UHPLC) are investigated and compared to high-pressure liquid chromatography (HPLC). The drug Omeprazole, together with three other model compounds (with different chemical characteristics, namely uncharged, positively and
Dennis Åsberg et al.
Journal of chromatography. A, 1362, 206-217 (2014-09-06)
This is the first investigation in a series that aims to enhance the scientific knowledge needed for reliable analytical method transfer between HPLC and UHPLC using the quality by design (QbD) framework. Here, we investigated the differences and similarities from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service