Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900686

Sigma-Aldrich

N-Acetylglycine

Vetec, reagent grade, 98%

Synonym(s):

Acetamidoacetic acid, Aceturic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2CO2H
CAS Number:
Molecular Weight:
117.10
Beilstein/REAXYS Number:
774114
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder or crystals

reaction suitability

reaction type: solution phase peptide synthesis

color

white

mp

207-209 °C (lit.)

SMILES string

CC(=O)NCC(O)=O

InChI

1S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)

InChI key

OKJIRPAQVSHGFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S K Mehta et al.
Journal of colloid and interface science, 314(2), 689-698 (2007-06-26)
The well-characterized, monodisperse nature of reverse micelles formed by sodium bis-(2-ethylhexyl)sulfosuccinate/water/isooctane and their usefulness in assimilating compounds of varied interests have been exploited to investigate the effect of acetyl modified amino acids (MAA) viz., N-acetyl-L-glycine (NAG), N-acetyl-L-aspartic acid (NAA) and
Functionalizing glycine derivatives by direct C-C bond formation.
Liang Zhao et al.
Angewandte Chemie (International ed. in English), 47(37), 7075-7078 (2008-08-02)
Bram Boeckx et al.
The journal of physical chemistry. A, 116(8), 1956-1965 (2012-01-26)
Within the structure determination task for peptides, which is of large interest due to the relation between structure and functionality, infrared spectra can provide detailed information on the conformational behavior. The conformational landscape ofN-acetylgycine has been studied by a combined
Liangxing Wu et al.
Journal of the American Chemical Society, 130(12), 4089-4096 (2008-03-07)
Eight B-containing compounds, i.e., 1a-h, were prepared as mimics of the green fluorescent protein (GFP) fluorophore. The underlying concept was that synthetic GFP chromophore analogues are not fluorescent primarily because of free rotation about an aryl-alkene bond (Figure 1b). This
H K Ogawa et al.
Journal of chromatography, 612(1), 145-149 (1993-01-29)
A simple and sensitive method for the determination of N-acetylneuraminic acid and N-glycolylneuraminic acid in glycoproteins by high-performance liquid chromatography has been investigated. The best condition to release sialic acid from glycoproteins involved incubation at 80 degrees C for 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service