Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900682

Sigma-Aldrich

4-Hydroxypiperidine

Vetec, reagent grade, 98%

Synonym(s):

4-Piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein/REAXYS Number:
102738
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

bp

108-114 °C/10 mmHg (lit.)

SMILES string

OC1CCNCC1

InChI

1S/C5H11NO/c7-5-1-3-6-4-2-5/h5-7H,1-4H2

Inchi Key

HDOWRFHMPULYOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service