Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900681

Sigma-Aldrich

3,5-Diaminobenzoic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(H2N)2C6H3CO2H
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
2086484
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

235-238 °C (dec.) (lit.)

SMILES string

Nc1cc(N)cc(c1)C(O)=O

InChI

1S/C7H8N2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,8-9H2,(H,10,11)

InChI key

UENRXLSRMCSUSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interference of Bis-Tris buffer with the diaminobenzoic acid fluorescence assay used to quantify DNA.
W E Schy et al.
Mutation research, 226(4), 263-266 (1989-08-01)
J Beckmann et al.
Acta diabetologica latina, 18(1), 51-57 (1981-01-01)
DNA content seems to be an ideal reference parameter for data on secretory function or metabolism of pancreatic islets. The approved fluorometric DNA assay with diaminobenzoic acid (DABA) of Kissane and Robins comprises repeated ethanol extractions of the tissue for
Céline Douat-Casassus et al.
Journal of the American Chemical Society, 126(25), 7817-7826 (2004-06-24)
Peptide dendrimers incorporating 3,5-diaminobenzoic acid 1 as a branching unit (B) were prepared by solid-phase synthesis of ((Ac-A(3))(2)B-A(2))(2)B-Cys-A(1)-NH(2) followed by disulfide bridge formation. Twenty-one homo- and heterodimeric dendrimers were obtained by permutations of aspartate, histidine, and serine at positions A(1)
[Improved fluorometric method for DNA microanalysis].
M M Vilenchik et al.
Izvestiia Akademii nauk SSSR. Seriia biologicheskaia, (2)(2), 217-222 (1984-03-01)
Wei-Yu Chen et al.
Analytical chemistry, 75(16), 4223-4228 (2003-11-25)
The alkali cation adductions of oligonucleotides dramatically degrade MALDI mass spectra and even affect the detection limit. Desalting is generally involved in MALDI sample preparation. This work demonstrates the feasibility of using 3,4-diaminobenzoic acid (DABA) and 3,5-DABA as the MALDI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service