Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900678

Sigma-Aldrich

4′-Methylpropiophenone

Vetec, reagent grade, 96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4COC2H5
CAS Number:
Molecular Weight:
148.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

96%

refractive index

n20/D 1.528 (lit.)

bp

238-239 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

CCC(=O)c1ccc(C)cc1

InChI

1S/C10H12O/c1-3-10(11)9-6-4-8(2)5-7-9/h4-7H,3H2,1-2H3

InChI key

PATYHUUYADUHQS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

228.2 °F - closed cup

flash_point_c

109 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service