Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900677

Sigma-Aldrich

1,2,4-Benzenetriol

Vetec, reagent grade, 98%

Synonym(s):

Hydroxyhydroquinone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

reagent grade

product line

Vetec

assay

98%

mp

140 °C (subl.) (lit.)

SMILES string

Oc1ccc(O)c(O)c1

InChI

1S/C6H6O3/c7-4-1-2-5(8)6(9)3-4/h1-3,7-9H

InChI key

GGNQRNBDZQJCCN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Veloso Goulart et al.
Environmental monitoring and assessment, 192(12), 797-797 (2020-11-29)
Fipronil and 2,4-D are two main pesticides of sugarcane cultivation in Brazil. While pesticides have helped to improve food quality and quantity, the continuous increase in usage has raised concerns over the unintended negative environmental impacts they introduce. Mesocosm systems
Karolina Nordin et al.
Applied and environmental microbiology, 71(11), 6538-6544 (2005-11-05)
Arthrobacter chlorophenolicus A6, a previously described 4-chlorophenol-degrading strain, was found to degrade 4-chlorophenol via hydroxyquinol, which is a novel route for aerobic microbial degradation of this compound. In addition, 10 open reading frames exhibiting sequence similarity to genes encoding enzymes
Rolis Chien-Wei Hou et al.
Journal of biomedical science, 13(1), 89-99 (2005-11-26)
Hydroxyhydroquinone or 1,2,4-benzenetriol (BT) detected in the beverages has a structure that coincides with the water-soluble form of a sesame lignan, sesamol. We previously showed that sesame antioxidants had neuroprotective abilities due to their antioxidant properties and/or inducible nitric oxide
Tohru Yamaguchi et al.
Nutrition, metabolism, and cardiovascular diseases : NMCD, 18(6), 408-414 (2007-10-24)
Coffee is rich in chlorogenic acids (CGA), whose metabolites may have beneficial effects such as anti-hypertensive effects. However, trial results concerning the effects of coffee on blood pressure (BP) are not consistent. A recent study suggested that hydroxyhydroquinone (HHQ), produced
Atsushi Suzuki et al.
FEBS letters, 580(9), 2317-2322 (2006-04-04)
Chlorogenic acid, a polyphenol found in coffee, has antihypertensive actions, but epidemiologic data on the effects of coffee on blood pressure are controversial. Specific coffee components that inhibit the hypotensive effect of chlorogenic acid and the physiologic mechanisms underlying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service