Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900676

Sigma-Aldrich

Sesamol

Vetec, reagent grade, 98%

Synonym(s):

3,4-(Methylenedioxy)phenol, 5-Benzodioxolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
Beilstein/REAXYS Number:
127405
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

mp

62-65 °C (lit.)

SMILES string

Oc1ccc2OCOc2c1

InChI

1S/C7H6O3/c8-5-1-2-6-7(3-5)10-4-9-6/h1-3,8H,4H2

InChI key

LUSZGTFNYDARNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Chien Chang et al.
Journal of biomedical science, 18, 93-93 (2011-12-16)
Platelet activation is relevant to a variety of coronary heart diseases. Our previous studies revealed that sesamol possesses potent antiplatelet activity through increasing cyclic AMP formation. Although platelets are anucleated cells, they also express the transcription factor, NF-κB, that may
Ashok Kumar Sharma et al.
The Journal of nutritional biochemistry, 23(11), 1482-1489 (2012-03-09)
Increased oxidative stress and inflammation in obesity are the central and causal components in the pathogenesis and progression of cardiometabolic syndrome (CMetS). The aim of the study was to determine the potential role of sesamol (a natural powerful antioxidant and
Soheila Kashanian et al.
Molecular biology reports, 40(2), 1173-1179 (2012-10-17)
The interaction of native calf thymus DNA (CT-DNA) with sesamol (3,4-methylenedioxyphenol) in Tris-HCl buffer at neutral pH 7.4 was monitored by absorption spectrophotometry, viscometry and spectrofluorometry. It is found that sesamol molecules could interact with DNA outside and/or groove binding
Chifumi Kinugasa et al.
Blood coagulation & fibrinolysis : an international journal in haemostasis and thrombosis, 22(6), 526-531 (2011-05-18)
Prevention of arterial thrombotic diseases has a high priority in developed countries. An inappropriate diet is known to enhance the risks for acute thrombotic events, and nutritional products experimentally shown to be antithrombotic, might contribute beneficial effects. The present study
Srinivasan Periasamy et al.
Cell biochemistry and biophysics, 61(2), 327-336 (2011-06-18)
We investigated the therapeutic effect of sesamol against monocrotaline-induced sinusoidal obstruction syndrome (SOS) in rats. Male Sprague-Dawley rats were gavaged with a single dose of monocrotaline (90 mg/kg) to induce SOS. Sesamol (5, 10, 20, and 40 mg/kg) was subcutaneously

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service