Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

V900674

Sigma-Aldrich

2-Thiophenecarboxylic acid

Vetec, reagent grade, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4O2S
CAS Number:
Molecular Weight:
128.15
Beilstein/REAXYS Number:
110150
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

98%

bp

260 °C (lit.)

mp

125-127 °C (lit.)

SMILES string

OC(=O)c1cccs1

InChI

1S/C5H4O2S/c6-5(7)4-2-1-3-8-4/h1-3H,(H,6,7)

InChI key

QERYCTSHXKAMIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Savarin et al.
Organic letters, 3(14), 2149-2152 (2001-07-07)
[reaction: see text] A new protocol for the palladium-catalyzed, copper-mediated coupling of aryl and alkenyl iodides with boronic acids is described. As an alternative to the well-known and widely used Suzuki cross-coupling, this reaction occurs in the absence of a
Laval Chan et al.
Bioorganic & medicinal chemistry letters, 14(3), 797-800 (2004-01-27)
Further SAR studies on the thiophene-2-carboxylic acids are reported. These studies led to the identification of a series of tertiary amides that show inhibition of both HCV NS5B polymerase in vitro and HCV subgenomic RNA replication in Huh-7 cells. Structural
L G Raisz et al.
Calcified tissue international, 37(5), 556-559 (1985-09-01)
We have compared the effects of thiophene 2-carboxylic acid (TCA) and a number of sulfur- and nitrogen-containing analogs for their ability to inhibit bone resorption in organ cultures of fetal rat long bones. Four compounds,--thionaphthene-2-carboxylic acid (TNCA), dibenzo-thiophene-4-carboxylic acid, indole-2-carboxylic
Hao Li et al.
Organic letters, 13(14), 3682-3685 (2011-06-17)
A stereocontrolled synthesis of α-amino-α'-alkoxy ketones is described. This pH-neutral copper(I) thiophene-2-carboxylate (CuTC)-catalyzed cross-coupling of amino acid thiol esters and chiral nonracemic α-alkoxyalkylstannanes gives α-amino-α'-alkoxy ketones in good to excellent yields with complete retention of configuration at the α-amino- and
Laval Chan et al.
Bioorganic & medicinal chemistry letters, 14(3), 793-796 (2004-01-27)
The discovery of a novel class of HCV NS5B polymerase inhibitors, 3-arylsulfonylamino-5-phenyl-thiophene-2-carboxylic acids is described. SAR studies have yielded several potent inhibitors of HCV polymerase as well as of HCV subgenomic RNA replication in Huh-7 cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service