Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V900672

Sigma-Aldrich

3-(2-Hydroxyethyl)indole

Vetec, reagent grade, 97%

Synonym(s):

2-(3-Indolyl)ethanol, 3-Indoleethanol, IEA, NSC 3884, Tryptophol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO
CAS Number:
Molecular Weight:
161.20
Beilstein/REAXYS Number:
125553
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

mp

56-59 °C (lit.)

SMILES string

OCCc1c[nH]c2ccccc12

InChI

1S/C10H11NO/c12-6-5-8-7-11-10-4-2-1-3-9(8)10/h1-4,7,11-12H,5-6H2

InChI key

MBBOMCVGYCRMEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Inhibitors of the C-terminal domain of RNA polymerase II and their antitumor activities
  • Anti-HIV-1 agents
  • Inhibitors of Protein-Protein Interactions
  • Partial agonists of the serotonin 5-HT1A receptor
  • Growth hormone secretagogues
  • Vascular endothelial growth factor (VEGF) inhibitors
  • A2B adenosine receptor ligands
  • Potential detoxification inhibitors of the crucifer phytoalexin brassinin
  • Inhibitors of interleukine 6
  • Dual binding site acetylcholinesterase inhibitors

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Kehraus et al.
Journal of natural products, 65(7), 1056-1058 (2002-07-27)
Geometricin A (1), together with the known compounds (7E,12E,18S,20Z)-variabilin (2), clathryimine A (3), tryptophol (4), and L-tryptophan, has been isolated from the methanol extract of the Australian sponge Luffariella geometrica. The structure of the new compound geometricin A (1) was
B Adam Lenhart et al.
G3 (Bethesda, Md.), 9(5), 1533-1544 (2019-03-14)
In fungi, filamentous growth is a major developmental transition that occurs in response to environmental cues. In diploid Saccharomyces cerevisiae, it is known as pseudohyphal growth and presumed to be a foraging mechanism. Rather than unicellular growth, multicellular filaments composed
Alessandro Palmieri et al.
Organic & biomolecular chemistry, 10(17), 3486-3493 (2012-03-22)
Reduction of ketosulfonyl indoles with sodium borohydride provides a ready entry to tryptophols in a regiocomplementary fashion with respect to the traditional oxirane ring-opening by indoles under Friedel-Crafts conditions. Compared to traditional β-ketosulfones, ketosulfonyl indoles show a peculiar behavior since
Atiqur Rahman et al.
Bioscience, biotechnology, and biochemistry, 74(11), 2202-2208 (2010-11-13)
Rhizobacteria isolated from wild dipterocarp saplings in Central Kalimantan, Indonesia, were subjected to Salkowski's reagent test, which is often used in detecting indolic substances. Among 69 isolates grown in a low-nitrogen medium supplemented with L-tryptophan (TRP), culture fluids of 29
Hexon Angel Contreras-Cornejo et al.
Plant physiology, 149(3), 1579-1592 (2009-01-30)
Trichoderma species belong to a class of free-living fungi beneficial to plants that are common in the rhizosphere. We investigated the role of auxin in regulating the growth and development of Arabidopsis (Arabidopsis thaliana) seedlings in response to inoculation with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service