Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

V900668

Sigma-Aldrich

Sodium benzenesulfonate

Vetec, reagent grade, 97%

Synonym(s):

Benzenesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SO3Na
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
3918459
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

reagent grade

product line

Vetec

assay

97%

density

1.124 g/mL at 25 °C (lit.)

SMILES string

[Na+].[O-]S(=O)(=O)c1ccccc1

InChI

1S/C6H6O3S.Na/c7-10(8,9)6-4-2-1-3-5-6;/h1-5H,(H,7,8,9);/q;+1/p-1

InChI key

MZSDGDXXBZSFTG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramasubbu Ramani et al.
Biomacromolecules, 9(5), 1390-1397 (2008-04-19)
We present lamellar self-assembly of cationic poly(L-histidine) (PLH) stoichiometrically complexed with an anionic surfactant, dodecyl benzenesulfonic acid (DBSA), which allows a stabilized conformation reminiscent of polyproline type II (PPII) left-handed helices. Such a conformation has no intrapeptide hydrogen bonds, and
Wenyi Huang et al.
ACS nano, 6(11), 10178-10185 (2012-10-27)
High-performance graphene nanopapers are prepared from an aqueous solution of functional graphenes with benzenesulfonic acid groups via covalent bonds. The formed hydrophobic graphene nanopapers showed the highest tensile strength of 360 MPa and Young's modulus of 102 GPa for samples
Wen Huang et al.
The Journal of organic chemistry, 73(17), 6845-6848 (2008-08-07)
A novel, convenient, and efficient method has been developed for selective synthesis of spirocycle, polycycle, and di- and tetrahydronaphthalene systems from aryl-substituted propargylic alcohols by FeCl3- or TsOH-catalyzed multiple activations of unsaturated C-C bonds and C-H bonds.
Lekha Gupta et al.
The Journal of organic chemistry, 75(24), 8392-8399 (2010-11-17)
A combination of NMR, kinetic, and computational methods are used to examine reactions of lithium diethylamide in tetrahydrofuran (THF) with n-dodecyl bromide and n-octyl benzenesulfonate. The alkyl bromide undergoes competitive S(N)2 substitution and E2 elimination in proportions independent of all
Erzsébet Szabó-Bárdos et al.
Water research, 45(4), 1617-1628 (2010-12-28)
Titanium dioxide-mediated photocatalyzed degradation of benzenesulfonate (BS) was investigated by monitoring chemical oxygen demand (COD), total organic carbon (TOC) content, sulfate concentration, pH as well as the absorption and emission spectral changes in both argon-saturated and aerated systems. Liquid chromatography-mass

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service